Gene Transcripts

Gene Symbol
AP3B2
Entrez Gene ID
8120
Gene Name
adaptor related protein complex 3 beta 2 subunit
Gene Aliases
NAPTB
Chromosome Location
Chr. 15: 82659281 - 82710112 on Build GRCh38
UniGene ID
Hs.199593
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278511.1 NP_001265440.1 6 752
NM_001278512.1 NP_001265441.1 7 848
NM_004644.4 NP_004635.2 7 848
XM_017022640.1 XP_016878129.1 7 1052
XM_017022641.1 XP_016878130.1 7 1052
GenBank mRNA AF022152.1 - 7 677
BC026700.1 - 7 800
BC093739.1 - 7 719 7939584
BC143342.1 - 6 623 9051847
BC143346.1 - 7 719 9051851
CR749860.1 - 6 848
U37673.1 - 7 707

View Full Product Details