Gene Transcripts

Gene Symbol
Limch1
Entrez Gene ID
305332
Gene Name
LIM and calponin homology domains 1
Gene Aliases
RGD1305269
Chromosome Location
Chr. 14: 42760363 - 43072843 on Build Rnor_6.0
UniGene ID
Rn.232004
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191678.1 NP_001178607.1 3 198
XM_008770147.1 XP_008768369.1 3 1771

View Full Product Details