Gene Transcripts

Gene Symbol
Tapt1
Entrez Gene ID
305386
Gene Name
transmembrane anterior posterior transformation 1
Gene Aliases
RGD1309656
Chromosome Location
Chr. 14: 71416153 - 71462646 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770236.1 XP_008768458.1 5 800

View Full Product Details