Gene Transcripts

Gene Symbol
SYMPK
Entrez Gene ID
8189
Gene Name
symplekin
Gene Aliases
SPK, SYM
Chromosome Location
Chr. 19: 45815442 - 45863290 on Build GRCh38
UniGene ID
Hs.515475
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004819.2 NP_004810.2 4 430
XM_005259286.1 XP_005259343.1 4 383
XM_011527354.1 XP_011525656.1 5 433
GenBank mRNA AB209024.1 - 4 2678
AB210010.1 - 4 275
AF049526.1 - 4 220
AK130602.1 - 4 287
AK300252.1 - 4 284
BC006536.2 - 3 137
BC006567.2 - 3 137
BC030214.1 - 4 272
BX649165.1 - 4 285
DB044963.1 - 4 406
DB461873.1 - 4 430
U88726.1 - 3 201
Y10931.1 - 4 249

View Full Product Details