Gene Transcripts
- Gene Symbol
- SYMPK
- Entrez Gene ID
- 8189
- Gene Name
- symplekin
- Gene Aliases
- SPK, SYM
- Chromosome Location
- Chr. 19: 45815442 - 45863290 on Build GRCh38
- UniGene ID
- Hs.515475
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004819.2 |
NP_004810.2
|
15, 16 |
2314 |
|
XM_005259286.1 |
XP_005259343.1
|
15, 16 |
2267 |
|
XM_011527354.1 |
XP_011525656.1
|
16, 17 |
2317 |
|
GenBank mRNA |
AB209024.1 |
- |
15, 16 |
4477 |
|
AB210010.1 |
- |
15, 16 |
2159 |
|
AK300252.1 |
- |
15, 16 |
2168 |
|
BC006536.2 |
- |
14, 15 |
2021 |
|
BC006567.2 |
- |
14, 15 |
2021 |
|
BX649165.1 |
- |
14, 15 |
3220 |
|
U49240.1 |
- |
10, 11 |
1783 |
|
Y10931.1 |
- |
15, 16 |
2132 |
|
View Full Product Details