Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006767.3 | NP_006758.2 | 14 | 1701 | |
GenBank mRNA | AK296347.1 | - | 13 | 1582 | |
AK310747.1 | - | 12 | 1304 | ||
AK313502.1 | - | 9 | 1103 | ||
BC026214.1 | - | 14 | 1603 | 4399716 | |
CT841521.1 | - | 14 | 1617 | ||
D38496.1 | - | 14 | 1596 |