Gene Transcripts

Gene Symbol
Rgr
Entrez Gene ID
306307
Gene Name
retinal G protein coupled receptor
Gene Aliases
Chromosome Location
Chr. 16: 14286323 - 14300951 on Build Rnor_6.0
UniGene ID
Rn.26828
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107299.1 NP_001100769.1 5, 6 674
XM_008771101.1 XP_008769323.1 5, 6 662

View Full Product Details