Gene Transcripts

Gene Symbol
Snx25
Entrez Gene ID
306471
Gene Name
sorting nexin 25
Gene Aliases
Chromosome Location
Chr. 16: 49329136 - 49432399 on Build Rnor_6.0
UniGene ID
Rn.16962
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006253168.1 XP_006253230.1 4 518
XM_224863.6 XP_224863.5 4 518

View Full Product Details