Gene Transcripts

Gene Symbol
Lonrf1
Entrez Gene ID
306505
Gene Name
LON peptidase N-terminal domain and ring finger 1
Gene Aliases
RGD1562583
Chromosome Location
Chr. 16: 59536248 - 59594251 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771313.1 XP_008769535.1 9 1875

View Full Product Details