Gene Transcripts

Gene Symbol
Ror2
Entrez Gene ID
306782
Gene Name
receptor tyrosine kinase-like orphan receptor 2
Gene Aliases
Chromosome Location
Chr. 17: 11953552 - 12134386 on Build Rnor_6.0
UniGene ID
Rn.211583
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107339.1 NP_001100809.1 8 1486
XM_008771495.1 XP_008769717.1 7 1198

View Full Product Details