Gene Transcripts

Gene Symbol
Armc4
Entrez Gene ID
307036
Gene Name
armadillo repeat containing 4
Gene Aliases
ddx5-ps1
Chromosome Location
Chr. 17: 60283385 - 60474289 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006254139.2 XP_006254201.1 20 3653
XM_008771806.1 XP_008770028.1 19 3175
XM_008771807.1 XP_008770029.1 19 3106
XM_008771810.1 XP_008770032.1 19 3175

View Full Product Details