Gene Transcripts

Gene Symbol
Dip2c
Entrez Gene ID
307067
Gene Name
DIP2 disco-interacting protein 2 homolog C (Drosophila)
Gene Aliases
RGD1560155
Chromosome Location
Chr. 17: 63427016 - 63810183 on Build Rnor_6.0
UniGene ID
Rn.219346
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107360.1 NP_001100830.1 27 3457
XM_008771732.1 XP_008769954.1 27 3325

View Full Product Details