Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001244580.1 | NP_001231509.1 | 37 | 5335 | |
NM_003496.3 | NP_003487.1 | 36 | 5281 | ||
GenBank mRNA | AB209489.1 | - | 28 | 4291 | |
AF076974.1 | - | 36 | 5281 | ||
AF110377.1 | - | 37 | 5255 |