Gene Transcripts

Gene Symbol
Gan
Entrez Gene ID
307893
Gene Name
gigaxonin
Gene Aliases
Chromosome Location
Chr. 19: 49694779 - 49746178 on Build Rnor_6.0
UniGene ID
Rn.218497
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107434.1 NP_001100904.1 9, 10 1611
XM_006255685.2 XP_006255747.1 9, 10 2393

View Full Product Details