Gene Transcripts

Gene Symbol
Zbtb2
Entrez Gene ID
308126
Gene Name
zinc finger and BTB domain containing 2
Gene Aliases
Chromosome Location
Chr. 1: 40914175 - 40933401 on Build Rnor_6.0
UniGene ID
Rn.36118
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107460.1 NP_001100930.1 2 620
XM_008758705.1 XP_008756927.1 2 619

View Full Product Details