Gene Transcripts

Gene Symbol
Vom2r4
Entrez Gene ID
308248
Gene Name
vomeronasal 2 receptor, 4
Gene Aliases
RGD1564110
Chromosome Location
Chr. 1: 714270 - 719690 on Build Rnor_6.0
UniGene ID
Rn.217649
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099458.1 NP_001092928.1 746

View Full Product Details