Gene Transcripts

Gene Symbol
Pnmal2
Entrez Gene ID
308393
Gene Name
paraneoplastic Ma antigen family-like 2
Gene Aliases
RGD1560435
Chromosome Location
Chr. 1: 78876205 - 78878779 on Build Rnor_6.0
UniGene ID
Rn.98225
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107481.1 NP_001100951.1 1 1179

View Full Product Details