Gene Transcripts

Gene Symbol
Kctd14
Entrez Gene ID
308836
Gene Name
potassium channel tetramerization domain containing 14
Gene Aliases
Chromosome Location
Chr. 1: 162418413 - 162424322 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229795.2 XP_006229857.1 2 1640

View Full Product Details