Gene Transcripts

Gene Symbol
Mrpl21
Entrez Gene ID
309140
Gene Name
mitochondrial ribosomal protein L21
Gene Aliases
Chromosome Location
Chr. 1: 218532045 - 218545424 on Build Rnor_6.0
UniGene ID
Rn.22778
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107567.1 NP_001101037.1 4 512
XM_008760120.1 XP_008758342.1 4 414

View Full Product Details