Gene Transcripts

Gene Symbol
Mamdc2
Entrez Gene ID
309410
Gene Name
MAM domain containing 2
Gene Aliases
Chromosome Location
Chr. 1: 241071110 - 241223814 on Build Rnor_6.0
UniGene ID
Rn.23790
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760316.1 XP_008758538.1 9 1869
GenBank mRNA BC079474.1 - 3 223

View Full Product Details