Gene Transcripts

Gene Symbol
RT1-A3
Entrez Gene ID
309627
Gene Name
RT1 class I, locus A3
Gene Aliases
RT1-A, RT1.A3
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.116787
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008830.1 NP_001008830.1 3 255

View Full Product Details