Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001107615.1 | NP_001101085.1 | 1 | 205 | |
XM_006256129.2 | XP_006256191.1 | 2 | 645 | ||
XM_006256130.2 | XP_006256192.1 | 2 | 645 | ||
XM_006256131.2 | XP_006256193.1 | 2 | 354 | ||
XM_006256132.2 | XP_006256194.1 | 2 | 562 |