Gene Transcripts

Gene Symbol
Ankrd33b
Entrez Gene ID
310200
Gene Name
ankyrin repeat domain 33B
Gene Aliases
RGD1564227
Chromosome Location
Chr. 2: 84361061 - 84436912 on Build Rnor_6.0
UniGene ID
Rn.205946
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191565.1 NP_001178494.1 3 534
GenBank mRNA FQ232932.1 - 3 723

View Full Product Details