Gene Transcripts

Gene Symbol
Eif5a2
Entrez Gene ID
310261
Gene Name
eukaryotic translation initiation factor 5A2
Gene Aliases
eIF5AII
Chromosome Location
Chr. 2: 115336378 - 115354203 on Build Rnor_6.0
UniGene ID
Rn.47334
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100697.2 NP_001094167.1 4 486
XM_006232231.2 XP_006232293.1 4 1517

View Full Product Details