Gene Transcripts

Gene Symbol
Flg2
Entrez Gene ID
310586
Gene Name
filaggrin family member 2
Gene Aliases
RGD1566121
Chromosome Location
Chr. 2: 193483619 - 193485463 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008761273.1 XP_008759495.1 2 365

View Full Product Details