Gene Transcripts

Gene Symbol
Cd101
Entrez Gene ID
310727
Gene Name
CD101 molecule
Gene Aliases
Igsf2
Chromosome Location
Chr. 2: 203376012 - 203413895 on Build Rnor_6.0
UniGene ID
Rn.229951
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271217.1 NP_001258146.1 6 1721
XM_006233049.2 XP_006233111.1 5 1306
XM_008761362.1 XP_008759584.1 6 2491

View Full Product Details