Gene Transcripts

Gene Symbol
Ssna1
Entrez Gene ID
311802
Gene Name
Sjogren syndrome nuclear autoantigen 1
Gene Aliases
Chromosome Location
Chr. 3: 2489038 - 2490476 on Build Rnor_6.0
UniGene ID
Rn.145066
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006233613.1 XP_006233675.1 2 327
GenBank mRNA BC166994.1 - 2 243
CB576781.1 - 2 280
FN805842.1 - 2 222

View Full Product Details