Gene Transcripts

Gene Symbol
Zbtb26
Entrez Gene ID
311910
Gene Name
zinc finger and BTB domain containing 26
Gene Aliases
Chromosome Location
Chr. 3: 21685339 - 21697945 on Build Rnor_6.0
UniGene ID
Rn.218409
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107840.1 NP_001101310.1 2 634
XM_006234082.2 XP_006234144.1 1 506

View Full Product Details