Gene Transcripts

Gene Symbol
Lrig1
Entrez Gene ID
312574
Gene Name
leucine-rich repeats and immunoglobulin-like domains 1
Gene Aliases
Chromosome Location
Chr. 4: 126615998 - 126717439 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008763142.1 XP_008761364.1 5 1410
XM_232237.8 XP_232237.6 5 622

View Full Product Details