Gene Transcripts

Gene Symbol
Cyld
Entrez Gene ID
312937
Gene Name
cylindromatosis (turban tumor syndrome)
Gene Aliases
LRRGT00003, Rp1, Rp1h
Chromosome Location
Chr. 19: 19260506 - 19323817 on Build Rnor_6.0
UniGene ID
Rn.128760
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017380.1 NP_001017380.1 13 2397
XM_008772408.1 XP_008770630.1 13 3646
GenBank mRNA BC082001.1 - 13 2397

View Full Product Details