Gene Transcripts

Gene Symbol
Frmpd1
Entrez Gene ID
313244
Gene Name
FERM and PDZ domain containing 1
Gene Aliases
Chromosome Location
Chr. 5: 60752372 - 60837773 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006238143.2 XP_006238205.1 16 3912
XM_006238144.2 XP_006238206.1 16 3865
XM_008763690.1 XP_008761912.1 16 2620

View Full Product Details