Gene Transcripts

Gene Symbol
Plch2
Entrez Gene ID
313756
Gene Name
phospholipase C, eta 2
Gene Aliases
RGD1304675
Chromosome Location
Chr. 5: 172386387 - 172444477 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008764393.1 XP_008762615.1 9 2368
XM_008764394.1 XP_008762616.1 9 1425
XM_008764395.1 XP_008762617.1 9 1263
XM_008764396.1 XP_008762618.1 9 1416
XM_008764397.1 XP_008762619.1 10 1439
XM_008764398.1 XP_008762620.1 9 2367

View Full Product Details