Gene Transcripts

Gene Symbol
Thada
Entrez Gene ID
313865
Gene Name
thyroid adenoma associated
Gene Aliases
RGD1560519
Chromosome Location
Chr. 6: 7423555 - 7741433 on Build Rnor_6.0
UniGene ID
Rn.222825
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191769.1 NP_001178698.1 15 2337
XM_006239705.2 XP_006239767.1 16 2560
XM_006239707.2 XP_006239769.1 16 2560

View Full Product Details