Gene Transcripts

Gene Symbol
ARSB
Entrez Gene ID
411
Gene Name
arylsulfatase B
Gene Aliases
ASB, G4S, MPS6
Chromosome Location
Chr. 5: 78777209 - 78986534 on Build GRCh38
UniGene ID
Hs.149103
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000046.3 NP_000037.2 6 2469
NM_198709.2 NP_942002.1 7 1538
XM_011543390.1 XP_011541692.1 7 1682
XM_011543391.2 XP_011541693.1 6 1245
XM_011543392.2 XP_011541694.1 6 1245
XM_017009471.1 XP_016864960.1 6 1245
GenBank mRNA AK290865.1 - 6 1245
AK314903.1 - 6 1254
BC029051.1 - 7 1538 5186657
M32373.1 - 6 1742

View Full Product Details