Gene Transcripts

Gene Symbol
Utp20
Entrez Gene ID
314713
Gene Name
UTP20, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases
RGD1560606
Chromosome Location
Chr. 7: 29296912 - 29378105 on Build Rnor_6.0
UniGene ID
Rn.162943
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191779.1 NP_001178708.1 8 767
XM_006241227.2 XP_006241289.1 8 952

View Full Product Details