Gene Transcripts

Gene Symbol
NOP14
Entrez Gene ID
8602
Gene Name
NOP14 nucleolar protein
Gene Aliases
C4orf9, NOL14, RES4-25, RES425, UTP2
Chromosome Location
Chr. 4: 2937936 - 2963506 on Build GRCh38
UniGene ID
Hs.627133
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291978.1 NP_001278907.1 7 1154
NM_001291979.1 NP_001278908.1 7 1154
NM_003703.2 NP_003694.1 7 1154
GenBank mRNA AB000467.1 - 5 550
AF040965.1 - 7 1016
AK025692.1 - 1 19
AK292939.1 - 7 1154
BC009760.2 - 3 348
BC026035.1 - 7 981 4878317
BC053635.1 - 7 1033 5114280

View Full Product Details