Gene Transcripts
- Gene Symbol
- Naprt1
- Entrez Gene ID
- 315085
- Gene Name
- nicotinate phosphoribosyltransferase domain containing 1
- Gene Aliases
- Naprt
- Chromosome Location
- Chr. 7: 116922979 - 116926436 on Build Rnor_6.0
- UniGene ID
- Rn.22665
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207609.1 |
NP_997492.1
|
10 |
1358 |
|
GenBank mRNA |
AY214330.1 |
- |
10 |
1358 |
|
View Full Product Details