Gene Transcripts

Gene Symbol
Nckap1l
Entrez Gene ID
315348
Gene Name
NCK associated protein 1 like
Gene Aliases
Hem1
Chromosome Location
Chr. 7: 145068237 - 145113507 on Build Rnor_6.0
UniGene ID
Rn.149002
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108119.1 NP_001101589.1 13 1360
XM_008765753.1 XP_008763975.1 13 1416

View Full Product Details