Gene Transcripts
- Gene Symbol
- CDK13
- Entrez Gene ID
- 8621
- Gene Name
- cyclin dependent kinase 13
- Gene Aliases
- CDC2L, CDC2L5, CHED, hCDK13
- Chromosome Location
- Chr. 7: 39949501 - 40097134 on Build GRCh38
- UniGene ID
- Hs.233552
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003718.4 |
NP_003709.3
|
2, 3 |
2154 |
|
NM_031267.3 |
NP_112557.2
|
2, 3 |
2154 |
|
XM_011515597.2 |
XP_011513899.1
|
2, 3 |
3013 |
|
XM_017012750.1 |
XP_016868239.1
|
2, 3 |
3013 |
|
XM_017012751.1 |
XP_016868240.1
|
2, 3 |
3013 |
|
GenBank mRNA |
AJ297709.1 |
- |
2, 3 |
2004 |
|
AJ297710.1 |
- |
2, 3 |
2004 |
|
AK092534.1 |
- |
2, 3 |
2154 |
|
BC001274.1 |
- |
1, 2 |
424 |
|
BC080601.1 |
- |
2, 3 |
2477 |
|
M80629.1 |
- |
2, 3 |
801 |
|
View Full Product Details