Gene Transcripts

Gene Symbol
Sema7a
Entrez Gene ID
315711
Gene Name
semaphorin 7A, GPI membrane anchor
Gene Aliases
Chromosome Location
Chr. 8: 62723788 - 62746536 on Build Rnor_6.0
UniGene ID
Rn.214059
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108153.1 NP_001101623.1 10 1272
XM_008766273.1 XP_008764495.1 10 1119

View Full Product Details