Gene Transcripts

Gene Symbol
Fam212a
Entrez Gene ID
316001
Gene Name
family with sequence similarity 212, member A
Gene Aliases
RGD1560778
Chromosome Location
Chr. 8: 116763149 - 116764839 on Build Rnor_6.0
UniGene ID
Rn.35964
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108187.1 NP_001101657.1 2 570
GenBank mRNA FQ215477.1 - 2 564

View Full Product Details