Gene Transcripts

Gene Symbol
Rtp3
Entrez Gene ID
316018
Gene Name
receptor (chemosensory) transporter protein 3
Gene Aliases
Tmem7
Chromosome Location
Chr. 8: 119260609 - 119265475 on Build Rnor_6.0
UniGene ID
Rn.198821
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108190.1 NP_001101660.1 3 887
XM_006243965.2 XP_006244027.1 3 1221

View Full Product Details