Gene Transcripts

Gene Symbol
Lama1
Entrez Gene ID
316758
Gene Name
laminin, alpha 1
Gene Aliases
Chromosome Location
Chr. 9: 115916907 - 116042123 on Build Rnor_6.0
UniGene ID
Rn.2807
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108237.2 NP_001101707.2 34 5380
GenBank mRNA AB425340.1 - 34 5380
AB425341.1 - 34 5380

View Full Product Details