Gene Transcripts
- Gene Symbol
- Hhipl2
- Entrez Gene ID
- 317051
- Gene Name
- hedgehog interacting protein-like 2
- Gene Aliases
- RGD1311317
- Chromosome Location
- Chr. 13: 101814574 - 101834641 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_229754.7 |
XP_229754.4
|
4 |
1406 |
|
View Full Product Details