Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008773.2 | NP_001008773.2 | 3 | 692 | |
XM_006254688.2 | XP_006254750.1 | 3 | 809 | ||
XM_006254689.1 | XP_006254751.1 | 2 | 547 | ||
GenBank mRNA | AY325156.1 | - | 2 | 316 | |
BC093607.1 | - | 3 | 692 | 7318425 |