Gene Transcripts

Gene Symbol
Uprt
Entrez Gene ID
317237
Gene Name
uracil phosphoribosyltransferase (FUR1) homolog (S. cerevisiae)
Gene Aliases
RGD1564806
Chromosome Location
Chr. X: 75382567 - 75408539 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006257154.2 XP_006257216.1 5 739
XM_228538.8 XP_228538.3 6 691

View Full Product Details