Gene Transcripts

Gene Symbol
Tusc5
Entrez Gene ID
360576
Gene Name
tumor suppressor candidate 5
Gene Aliases
DSPB1
Chromosome Location
Chr. 10: 63471330 - 63491713 on Build Rnor_6.0
UniGene ID
Rn.18571
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039163.1 NP_001034252.1 2 914
GenBank mRNA AB218813.1 - 2 914

View Full Product Details