Gene Transcripts

Gene Symbol
Icam2
Entrez Gene ID
360647
Gene Name
intercellular adhesion molecule 2
Gene Aliases
Chromosome Location
Chr. 10: 94569889 - 94581211 on Build Rnor_6.0
UniGene ID
Rn.162206
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007725.1 NP_001007726.1 5 477
XM_006247624.2 XP_006247686.1 5 487
XM_006247627.2 XP_006247689.1 3 321
XM_008768377.1 XP_008766599.1 4 1084
XM_008768378.1 XP_008766600.1 6 536
XM_008768379.1 XP_008766601.1 4 396
GenBank mRNA BC079465.1 - 5 477
FQ220624.1 - 2 183
FQ231566.1 - 5 345
FQ231887.1 - 5 345

View Full Product Details