Gene Transcripts

Gene Symbol
Dnai2
Entrez Gene ID
360654
Gene Name
dynein, axonemal, intermediate chain 2
Gene Aliases
Dnaic2
Chromosome Location
Chr. 10: 103266343 - 103301521 on Build Rnor_6.0
UniGene ID
Rn.137787
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007726.1 NP_001007727.1 6 1049
GenBank mRNA BC081920.1 - 6 1049

View Full Product Details