Gene Transcripts

Gene Symbol
Pnpla6
Entrez Gene ID
360753
Gene Name
patatin-like phospholipase domain containing 6
Gene Aliases
RGD1564611
Chromosome Location
Chr. 12: 2068741 - 2098139 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008769004.1 XP_008767226.1 2 402
XM_008769005.1 XP_008767227.1 2 402
XM_008769006.1 XP_008767228.1 2 402
XM_008769007.1 XP_008767229.1 2 402
XM_008769008.1 XP_008767230.1 2 402
XM_008769009.1 XP_008767231.1 5 536
XM_008769010.1 XP_008767232.1 5 533
XM_008769011.1 XP_008767233.1 4 250
XM_008769012.1 XP_008767234.1 4 250
XM_008769013.1 XP_008767235.1 4 476
XM_008769014.1 XP_008767236.1 2 402
XM_008769015.1 XP_008767237.1 2 402

View Full Product Details